▸
Organic compounds
▸
Organic acids and derivatives
▸
Organic carbonic acids and derivatives
▸
Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™
Have Questions?
Change view
Quantity:
10 kg
500 g
Packaging:
View all
Poly Drum
Catalog number
FBRBP169-10
Price (MYR)
6,596.00
Each
Add to cart
Quantity:
10 kg
Packaging:
Poly Drum
Request bulk or custom format
Price (MYR)
6,596.00
Each
Add to cart
Specifications
Chemical Name or Material
Urea
CAS
57-13-6
ChemAlert Storage Symbol
Gray
Absorbance
0.05 max. (5M solution) at 280nm
Melting Point
131°C
View more
Urea is used as a denaturing agent for DNA and proteins.