▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Cleaners and Disinfectants
▸
Laboratory Surfactants and Wetting Agents
▸
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
For protein extraction procedures and SDS polyacrylamide gels | CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
Change view
Quantity:
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number
FBRBP166-100
Price (MYR)
490.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
490.00
Each
Add to cart
Specifications
Absorbance
0.1 max. (0.1% solution) at 280nm, 0.4 max. (0.1% solution) at 230nm
Chemical Name or Material
Sodium Dodecyl Sulfate
Color
White
CAS
151-21-3
ChemAlert Storage Symbol
Red
View more
For protein extraction procedures and SDS polyacrylamide gels
DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
SDS denatures proteins by disrupting the non-covalent bonds of the protein
Functionally tested for use in electrophoresis
Intended for research use only; not for human or animal diagnostic purposes