Check Order Status
Agarose (Low-Melting, Nucleic Acid Recovery/Molecular Biology Grade), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Agarose (Low-Melting, Nucleic Acid Recovery/Molecular Biology Grade), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FBRBP165-25
25 g
Catalog number
FBRBP165-25
Price (MYR)
924.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
924.00
EA
Add to cart
Specifications
Chemical Name or Material
Agarose
Name Note
Low-Melting, Nucleic Acid Recovery
Physical Form
Powder/Solid
CAS
39346-81-1
ChemAlert Storage Symbol
Gray
View more