Check Order Status
TEMED (Electrophoresis), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
TEMED (Electrophoresis), Fisher BioReagents
For polymerization of acrylamide/bis-acrylamide
Have Questions?
Change view
Quantity:
26 mL
130 mL
Packaging:
Amber Glass
Catalog number
FBRBP150-20
Price (MYR)
425.00
EA
Add to cart
Quantity:
26 mL
Packaging:
Amber Glass
Request bulk or custom format
Price (MYR)
425.00
EA
Add to cart
Specifications
Chemical Name or Material
TEMED
CAS
110-18-9
ChemAlert Storage Symbol
Red
Melting Point
-55°C
Color
Colorless
View more
TEMED is a commonly used catalyst for the polymerization of acrylamide/bis-acrylamide.