Check Order Status
Agar, Powder / Flakes, Fisher BioReagents™
Actual product may vary
Fisher Chemical
Agar, Powder / Flakes, Fisher BioReagents™
Used as a solidifying agent for culture media.
Have Questions?
Catalog Number
Quantity
Packaging Type
FBRBP1423-2
2000 g
Poly Pail
Catalog number
FBRBP1423-2
Price (MYR)
3,211.00
EA
Add to cart
Quantity:
2000 g
Packaging Type:
Poly Pail
Price (MYR)
3,211.00
EA
Add to cart
Used as a solidifying agent for culture media
Used as a solidfying agent to provide the solid medium to support the growth of microbes for cell culture applications
Functionally tested for promotion of growth of E. coli and S. aureus
Typical concentration is 1.5% agar (15g/L) in water
CAS: 9002-18-0
Molecular Weight: 336.337 g/mol
Chemical Formula: C14H24O9
Specifications
Form
Powder
Grade
Molecular genetics
Product Type
Agar
For Use With (Application)
Used for a solidifying agent for culture media
Packaging Type
Poly Pail
Quantity
2000 g