Agarose (Low-Melting, <1kb DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™
Agarose (Low-Melting, &lt;1kb DNA/RNA/Genetic Analysis Grade), Fisher BioReagents&trade;
Actual product may vary
Agarose (Low-Melting, &lt;1kb DNA/RNA/Genetic Analysis Grade), Fisher BioReagents&trade;
Agarose (Low-Melting, &lt;1kb DNA/RNA/Genetic Analysis Grade), Fisher BioReagents&trade;
Fisher Chemical
Agarose (Low-Melting, <1kb DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™
Agarose, C12H18O9, CAS Number-9012-36-6, 100g, Gray, Cream, DNase free, 0.15 max., Pass Test, 306.12, 35 deg.C max., 500g/cm2min., Genetic Analysis, Low-Melting, <1kb DNA/RNA, Poly Bottle, Powder/Solid, DNase- and RNase-Free, RT, 0.15% max. | CAS: 9012-36-6 | C12H18O9
Have Questions?
Catalog NumberQuantity
FBRBP1360-100100 g
Catalog number FBRBP1360-100
Price (MYR)
2,461.00
EA
Add to cart
Quantity:
100 g
Price (MYR)
2,461.00
EA
Add to cart
Agarose, C12H18O9, CAS Number-9012-36-6, 100g, Gray, Cream, DNase free, 0.15 max., Pass Test, 306.12, 35 deg.C max., 500g/cm2min., Genetic Analysis, Low-Melting, <1kb DNA/RNA, Poly Bottle, Powder/Solid, DNase- and RNase-Free, RT, 0.15% max.
Specifications
Chemical Name or MaterialAgarose
Name NoteLow-Melting, <1kb DNA/RNA
Physical FormPowder/Solid
CAS9012-36-6
ChemAlert Storage SymbolGray
Recommended StorageRT
ColorCream
Quantity100 g
DNaseDNase free
Electroendosmosis (EEO)0.15 max.
Gelation Temperature35°C max.
Gel Strength500g/cm2min.
PackagingPoly Bottle
Purity Grade NotesDNase- and RNase-Free
Molecular FormulaC12H18O9
Formula Weight306.12
GradeGenetic Analysis
Unit SizeEA