Check Order Status
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™
Have Questions?
Change view
Quantity:
500 g
100 g
Packaging:
Poly Bottle
Catalog number
FBRBP1356-500
Price (MYR)
2,919.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
2,919.00
EA
Add to cart
Specifications
Chemical Name or Material
Agarose
Name Note
Broad Separation Range for DNA/RNA (500bp to 25kb)
Physical Form
Powder/Solid
CAS
9012-36-6
ChemAlert Storage Symbol
Gray
View more
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade)