▸
Buffers and Standards
▸
Life Science Buffers
▸
TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP1335-500
500 mL
Poly Bottle
Catalog number
FBRBP1335-500
Price (MYR)
541.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
541.00
Each
Add to cart
Chemical Identifiers
CAS
64-19-7, 7732-18-5, 60-00-4, 77-86-1
Specifications
Chemical Name or Material
Tris-Acetate-EDTA
Name Note
10X Solution
CAS
64-19-7
ChemAlert Storage Symbol
Gray
CAS
7732-18-5
View more
Tris–Acetate–EDTA (TAE) is commonly used as a buffer for nucleic acid electrophoresis.