TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™
Fisher Chemical
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents™
CAS: 10043-35-3
Have Questions?
Catalog NumberQuantityPackaging
FBRBP1333-11 LNalgene Poly Bottle
Catalog number FBRBP1333-1
Price (MYR)
565.00
EA
Add to cart
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
Request bulk or custom format
Price (MYR)
565.00
EA
Add to cart
Specifications
Chemical Name or MaterialTris-Borate-EDTA
Name Note10X Solution
CAS10043-35-3
ChemAlert Storage SymbolGray
ColorColorless
View more
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis
Tris–Borate–EDTA (TBE) is commonly used as a buffer for nucleic acid electrophoresis.