Thermolyne™ Premium Large Muffle Furnaces
Actual product may vary
Thermo Scientific
Thermolyne™ Premium Large Muffle Furnaces
Conduct industrial applications, including ashing organic and inorganic samples and gravimetric analysis, with this premium large muffle furnace.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature ControlVoltageCertifications/Compliance
F6020CDig 1-setpoint w/ ramp/dwell (B1)240 VCSA approved
F6010Digital single setpoint (A1)240 VCSA approved
F6018Digital single setpoint (A1)208 VCSA approved
F6020C-33-80
also known as AVL
Digital with 4 programs (C1)240 VCSA approved, CE marked
F6020C-80Digital with 4 programs (C1)240 VCSA approved
F6030CM-33Digital with 4 programs, 16 segments240 VCSA approved, CE marked
Catalog number F6020C
HSN 85141900
Price (MYR)
45,452.00
Each
Temperature Control:
Dig 1-setpoint w/ ramp/dwell (B1)
Voltage:
240 V
Certifications/Compliance:
CSA approved
Price (MYR)
45,452.00
Each
Conduct industrial applications including ashing organic and inorganic samples and conducting gravimetric analysis with Thermo Scientific™ Thermolyne™ Premium Large Muffle Furnaces. Unit has four heating elements, which are located on the chamber top, bottom and sides for enhanced temperature uniformity. Built-in vent port removes undesirable contaminants and moisture to extend the life of the element and unit.
  • Four heating elements are located on the chamber top, bottom and sides for enhanced temperature uniformity
  • Built-in vent port removes undesirable contaminants and moisture to extend the life of the element and unit
  • Rear of the chamber incorporates a 0.95cm (0.38in.) dia. port for monitoring chamber temperatures with independent measuring devices
  • Optional stainless steel shelf doubles load capacity (maximum temperature of 900°C)
  • Door safety switch stops power to heating elements when door opens
  • Thermocouple break protection cuts power to the heating elements, preventing a thermocouple failure runaway condition
  • Furnaces with B1, C1, and D1 control also use a mechanical overtemperature protection relay
Choice of Temperature Controllers
  • Digital single setpoint control (A1): Single display shows actual temperature or setpoint
  • Digital single setpoint control with single ramp to setpoint and a dwell (B1): Single display shows actual temperature or setpoint
  • Digital programmable control with one stored program of eight segments (C1)
  • Digital programmable control with four stored programs, 16 segments per program (D1): Controller includes RS-232 communications
Includes:
Models F6010 and F6018 include a cord and plug set.
Requires:
All except Models F6010 and F6018 require a conduit connection.
Warranty: 1 year
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6020C
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeSingle Setpoint
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDig 1-setpoint w/ ramp/dwell (B1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A