MP-250 Series Digital Melting Point Apparatus
MP-250 Series Digital Melting Point Apparatus
Actual product may vary
Electrothermal
MP-250 Series Digital Melting Point Apparatus
This MP-250 Series Digital Melting Point Apparatus with rotating viewing head ensures each user can analyze samples clearly.
Have Questions?
Catalog NumberModelResolution
ELEIA9100Fixed Ramp Rate Model±0.1°C
Catalog number ELEIA9100
Price (MYR)
0.00
EA
Add to cart
Model:
Fixed Ramp Rate Model
Resolution:
±0.1°C
Price (MYR)
0.00
EA
Add to cart
Offers an easy way to measure the melting points of samples without sacrificing accuracy; the temperature resolution is within 0.1°C. Ideal for multiple users, being ergonomically designed to ensure comfortable viewing for everyone, with a height-adjustable extension arm and a viewing head that may be rotated to suit each user. Viewing head holds a viewing magnifier that offers 8X magnification.
  • View up to three samples simultaneously for greater efficiency.
  • Height-adjustable arm with viewing magnifier folds away for easy storage.
  • Integral light and 8x magnifying lens enhance sample observation.
  • Three audible beeps indicate when temperature is stable and ready for sample.
  • 4 x 1 melt memory capacity.
  • Adjustable object lens for sharp focus.
  • The MP-250D series digital melting point apparatus allows you to easily view three capillary tube samples simultaneously through the 8x magnifying lens.
  • The adjustable extension arm can be set to different viewing heights making it ideal for multiple users.
  • A rotating viewing head and safety eye piece eliminate glare on the lens and protect you against hot zone.
  • A 13-key membrane keypad offers easy-to-use menus.
  • The conveniently located push button controls allow temperature recording without looking away from the sample.
  • A digital microprocessor with ±0.1°C resolution provides fast warm-up and accurate temperature control.
  • Units can be calibrated by on site or by the manufacturer/service organization.
  • A calibration kit is required if you choose to calibrate on site.
Specifications
Product TypeDigital Melting Point Apparatus
Hertz50/60 Hz
Voltage100/240 V
Wattage45 W
Accuracy±1°C
Certifications/ComplianceCE
Dimensions (L x W x H)355 x 200 x 80 mm
Display TypeBacklit LCD
Electrical Requirements100/240 V, 50/60 Hz, 15 DC, 45 W
Height (English)17.75 in.
Height (Metric)45.1 cm
IncludesSupplied with a pack of 100 capillary tubes, power supply and power cable.
ModelFixed Ramp Rate Model
Resolution±0.1°C
Sensors100Ω Pt RTD
Temperature Range (English)Ambient to 752°F
Temperature Range (Metric)Ambient to 400°C
Weight (Metric)2.5 kg
Unit SizeEA