Barnstead™ Nanopure™ Accessories and Consumables
Actual product may vary
Thermo Scientific
Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Have Questions?
Catalog NumberType
D50256Pretreatment Cartridge Kit (Tap and DI Water)
Catalog number D50256
HSN 84212190
Price (MYR)
6,785.00
Each
Type:
Pretreatment Cartridge Kit (Tap and DI Water)
Price (MYR)
6,785.00
Each
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
  • Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
TypePretreatment Cartridge Kit (Tap and DI Water)
For Use With (Application)Water Purification
IncludesOne D50251 Tap/DI Pretreatment, three D50253 Ultrapure™ cartridges and two FL703X2 Remote Dispenser 0.2μm filters
Unit SizeEach