Barnstead™ Easypure™ Cartridges and Filters
Actual product may vary
Thermo Scientific
Barnstead™ Easypure™ Cartridges and Filters
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with replacement cartridges and filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)TypeFilterSizeResin Type
D502126Easypure II, Easypure UFCartridge Pack, Deionized Feed
D502124Easypure II UV, Easypure II UV/UFCartridge Pack, Deionized Feed
D50229EasyPure IICartridge and FilterUltrapure Mixed Bed
D50231EasyPure II CompactOrganic/Ion Exchange Pretreatment CartridgeActivated Carbon and Mixed Bed Ion Exchange Resin
D50233EasypureCleaning Cartridge
D502125Easypure II UV, Easypure II UV/UFCartridge Pack, RO or Distilled Feed
FL1192X1Easypure UF, Easypure II UV/UFUltrafilter
D50230Easypure IICartridge and Filter2/3 Macroreticular Resin
Catalog number D502126
HSN 84212190
Price (MYR)
2,829.00
Each
For Use With (Equipment):
Easypure II, Easypure UF
Type:
Cartridge Pack, Deionized Feed
Price (MYR)
2,829.00
Each
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.
D502126:
  • Includes: Organic/Colloid; two Thermo Scientific™ Ultrapure™ Ion Exchange
  • Type 1 filter
D502124:
  • Includes: Organic/Colloid, Ultrapure Ion Exchange, Organic Free Organic Removal
  • Ultralow filter with Organic Free Cartridge
D502127:
  • Includes: Organic/Ion Exchange, two Ultrapure Ion Exchange
  • Type 1 filter
D502125:
  • Includes: 09-034-18 Organic/Ion Exchange, 09-050-46 Ultrapure Ion Exchange, 09-034-16 Organic Free Organic Removal
  • Ultralow filter with Organic Free Cartridge
Specifications
Feedwater SourceDeionized
For Use With (Equipment)Easypure II, Easypure UF
TypeCartridge Pack, Deionized Feed
Unit SizeEach