Traceable™ Thermometer/Clock/Humidity Monitor
Actual product may vary
Traceable™ Thermometer/Clock/Humidity Monitor
Traceable™ Thermometer/Clock/Humidity Monitor
Traceable™ Thermometer/Clock/Humidity Monitor
Traceable™ Thermometer/Clock/Humidity Monitor
Traceable™ Thermometer/Clock/Humidity Monitor
Traceable™ Thermometer/Clock/Humidity Monitor
Traceable™ Thermometer/Clock/Humidity Monitor
Traceable™ Thermometer/Clock/Humidity Monitor
Traceable™ Thermometer/Clock/Humidity Monitor
Traceable™ Thermometer/Clock/Humidity Monitor
Traceable™ Thermometer/Clock/Humidity Monitor
Measures ambient temperature, relative humidity and tells time.
Have Questions?
Catalog NumberAlarmTypes
CTL06-662-4
Catalog number CTL06-662-4
Price (MYR)
374.00
Each
Price (MYR)
374.00
Each
Measures ambient temperature, relative humidity and tells time. Fisherbrand™ Traceable™ Thermometer/Clock/Humidity Monitors are designed for easy operation, accuracy and reliability in industrial, educational, hospital, and government labs. They are ideal for use in incubators, storerooms, drying chambers, hoods, cleanrooms, environmental cabinets, and general lab applications.
  • Three-line LCD display with 0.5 in.H (13mm) digits.
  • Dual ranges: 0° to 50°C and 32° to 122°F
  • Flip-down easel for desktop use
  • Switch selectable Celsius or Fahrenheit readings
  • Relative humidity range is 20 to 90%, resolution is 1% RH, and accuracy is ±8%
  • Memory recalls highest and lowest temperature and humidity readings
  • Clock shows time of day to the minute, with accuracy to 0.0190 seconds
  • Easy-to-read LCD reports temperature with accuracy to ±1°C and resolution to 0.1°C/°F
Specifications
TypeHygrometer
Humidity Range20 to 90%
Resolution0.1°C
Accuracy±1°C, ±5% (mid-range) to +±8% (end of range) RH
Temperature Range (Metric)0°C to 50°C
Temperature Accuracy±1°C
Display TypeLCD
Temperature Range (English)32°F to 122°F
Dimensions (L x W x H)4.25 x 2.25 x 0.5 in. (10.8 x 5.7 x 1.3 cm)
Length (English)4.25 in.
Length (Metric)10.8 cm
Width (English)2.25 in.
Width (Metric)5.7 cm
Height (English)0.5 in.
Height (Metric)1.3 cm
Weight (English)2.5 oz.
Weight (Metric)71 g
For Use With (Application)Incubator, storerooms, drying chambers, hoods, cleanrooms, environmental cabinets and general lab applications
Includes1.5V AAA battery, built-in stand and wall mount
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEach