ProChlor
ProChlor
Actual product may vary
ProChlor
Contec ProChlor is a revolutionary new sporicide achieving a 100% kill against spores in under 1 min.
Have Questions?
Catalog NumberContainer TypeQuantitySterility
CTCSBT102PCTrigger Spray Bottle6 x 1 LSterile
Catalog number CTCSBT102PC
Price (MYR)
2,301.00
CS
Add to cart
Container Type:
Trigger Spray Bottle
Quantity:
6 x 1 L
Sterility:
Sterile
Price (MYR)
2,301.00
CS
Add to cart
Ensures cleanroom is free from contamination and particulates.
A blend of hypochlorous acid in purified water, Contec ProChlor is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts, viruses and spores.
Contec ProChlor is 0.2 micron filtered, and filled in a Grade A environment. Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms.
Features & Benefits
  • Log 6 kill against Bacillus subtilis in 60 secs saves time spent on biodecontamination
  • Filtered to 0.2 microns in a Grade B cleanroom ensures the product is free from contamination and particulates
  • Sterile version available for Grade A and B cleanrooms
  • Contains no quaternary ammonium or surfactant so it's very low residue
  • No hazard classification means only basic PPE required and no special disposal is required
  • Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
  • Not classed as corrosive and can be used safely in all areas of the cleanroom
Note: For trigger spray option, see 15567161.
Specifications
Product TypeSporicide
FormLiquid Spray
pH3 to 6
SterilitySterile
AppearanceColorless
AutoclavableNot Autoclavable
ColorClear
CompositionStabilized Hypochlorous acid blended with purified water
Concentration2000 ppm hypochlorous acid
Container TypeTrigger Spray Bottle
DisposableLimited-use
FilteredFiltered
FlammableNo
For Use With (Application)Ready-to-Use Hard Surface Disinfectant
PackagingDouble bagged in polyethylene
Quantity6 x 1 L
Shelf Life2 Years
Volume (English)33.81 oz.
Volume (Metric)1 L
Unit SizeCS
Contents & Storage
Do not freeze. Store below 40°C.