PeridoxRTU
PeridoxRTU
Actual product may vary
PeridoxRTU
PeridoxRTU is a blend of hydrogen peroxide and peracetic acid.
Have Questions?
Catalog NumberVolume (Metric)
CTCSBC0902PX0.9 L
Catalog number CTCSBC0902PX
Price (MYR)
-
Volume (Metric):
0.9 L
  • Contec PeridoxRTU is a BPR and EPA authorised sporicide achieving greater than a log 3 reduction against bacterial and fungal spores in under 3 min
  • A blend of hydrogen peroxide and peracetic acid, Contec PeridoxRTU is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts and spores
  • PeridoxRTU's proprietary formulation contains surfactants for cleaning efficiency along with corrosion inhibitors for surface protection
  • PeridoxRTU can be used to clean and disinfect surfaces without damage or loss of functionality
  • Contec PeridoxRTU is filtered and filled in a controlled environment
  • The sterile product is provided validated sterile to a Sterility Assurance Level of 10-6
  • Provided double or single bagged, the product is designed for ease of entry into pharmaceutical cleanrooms
  • Available as either a 0.9 L bottle with a push/pull cap for small areas or in 3.78 L capped containers for mop head saturation or when larger volumes are required
  • Patented hyperactive chemistry makes it fast acting so save time spent of biodecontamination
  • Validated sterile version available makes it suitable for Grade A and B cleanrooms
  • Excellent material compatibility and will not damage common cleanroom materials
  • Double/single bagged helps facilitate transfer disinfection into cleanroom
  • Push/pull cap on 0.9 L eliminates droplets reducing the risk of inhalation and can be easily applied to a wipe or cleaning too
Specifications
Product TypeSporicide
FormLiquid
pHNot Applicable
SterilitySterile
AppearanceClear
AutoclavableNot Autoclavable
ColorClear
CompositionHydrogen Peroxide and Peracetic Acid
ConcentrationRTU
DisposableLimited-use
FilteredNon-filtered
FlammableNo
Volume (Metric)0.9 L
Unit SizeCS