EasyReach™ Cleaning Pads
Actual product may vary
Corning
EasyReach™ Cleaning Pads
EasyReach™ Cleaning Pads are very absorbent, which ensures even coverage of disinfectant or IPA.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)Product TypeMaterialSterility
CTCMEQT0002SZ7.5 x 2.8 in. (19 x 7 cm)Cleaning PadQuilted PolyesterSterile
CTCAMEC01137.5 x 2.8 in. (19 x 7 cm)Cleaning PadQuilted PolyesterSterile
Catalog number CTCMEQT0002SZ
Price (MYR)
2,494.00
Each
Dimensions (L x W):
7.5 x 2.8 in. (19 x 7 cm)
Product Type:
Cleaning Pad
Material:
Quilted Polyester
Sterility:
Sterile
Price (MYR)
2,494.00
Each
  • Single-use sealed polyester
  • Provide a low level of particulates which is essential for use in product contact areas
  • Very absorbent, ensuring good even coverage of disinfectant or IPA
  • Easy grab edge on the cleaning pad ensures the pad fits tightly on the mop frame
  • Sterile cleaning pads with dry and presaturated options
Specifications
Product TypeCleaning Pad
Length (English)7.5 in.
Length (Metric)19 cm
Width (English)2.8 in.
Width (Metric)7 cm
MaterialQuilted Polyester
AutoclavableNot Autoclavable
ColorWhite
DisposableSingle-use
Dimensions (L x W)7.5 x 2.8 in. (19 x 7 cm)
For Use With (Equipment)Cleaning Isolators, LAF Hoods, Glove Boxes and Other Mini Environments
Packaging Quantity4/Bag, 4 Pads/Bag, 15 Bags/Cs.