▸
Facility Safety and Maintenance
▸
Cleaning Supplies and Equipment
▸
Cleaners and Disinfectants
▸
Controlled Environment Cleaners and Disinfectants
▸
CyQuanol Non-Sterile Disinfectant Solution
Actual product may vary
Corning
CyQuanol Non-Sterile Disinfectant Solution
Contec™ CyQuanol is a blend of quaternary ammonium and ethyl alcohol that generates low residue.
Have Questions?
Catalog Number
Volume (English)
CTCFBC128CQ
1 gal.
Catalog number
CTCFBC128CQ
Price (MYR)
2,556.00
Case
Add to cart
Volume (English):
1 gal.
Price (MYR)
2,556.00
Case
Add to cart
0.2 micron filtered for the most critical environments
Bactericidal, virucidal, fungicidal and tuberculocidal
1-minute contact time makes it easier to achieve dwell time
No pre-clean required for this one-step cleaner disinfectant
Bleach-free makes it compatible with common cleanroom surfaces
Specifications
Product Type
Cleaner/Disinfectant
Sterility
Non-sterile
Appearance
Clear
Shelf Life
24 Months
Volume (English)
1 gal.
Volume (Metric)
3.8 L