Specifications
Product TypeSporicidal Disinfectant and Cleaner
FormLiquid
pH2.1 to 2.2 (at 20°C)
SterilitySterile
AppearanceWhite
Certifications/ComplianceBroad spectrum EPA-registered
Container TypeSingle bagged with re-closable cap
PackagingSingle bagged
Quantity6/Cs.
Shelf Life2 Year
Volume (English)1 qt.
Volume (Metric)946.3 mL
Unit SizeCase