Specifications
Product TypeCleaner and Disinfectant Solution
pH2 to 2.5
SterilityNon-sterile
CompositionHydrogen Peroxide
For Use With (Application)Cleaning and disinfecting environmental surfaces in cleanrooms and labs
Volume (English)32 oz.
Volume (Metric)0.9 L
Unit SizeCase