PREempt™ RTU Disinfectant Wipes
Actual product may vary
Corning
PREempt™ RTU Disinfectant Wipes
Clean and disinfect environmental surfaces in cleanrooms and laboratory areas including work stations, fume hoods, equipment and other hard non-porous environmental surfaces. Contec™ PREempt™ RTU Disinfectant Wipes helps to ensure user, protocol and product compliance with fast contact times and broad-spectrum efficacy.
Have Questions?
Catalog NumberDimensions (L x W)Packaging Quantity
CTC212216 x 7 in.12 x 160-Count Canister
Catalog number CTC21221
Price (MYR)
1,787.00
Case
Dimensions (L x W):
6 x 7 in.
Packaging Quantity:
12 x 160-Count Canister
Price (MYR)
1,787.00
Case
  • AHP - Utilizes AHP™, a patented synergistic blend of commonly used, safe ingredients that when combined with low levels of hydrogen peroxide dramatically increase potency and cleaning performance
  • Cleaner - Effective disinfectant as well as a cleaner resulting in added confidence that disinfection can occur
  • Faster - Rapid and realistic contact times on a broad spectrum of difficult to kill pathogens
  • Responsible - Designed to be easier on surfaces, staff and occupants. Users are not exposed to VOCs (volatile organic compounds) and PPE is not required as per health and safety rating
  • 5 minute tuberculocidal
  • 10 minute fungicidal
  • 30 seconds broad-spectrum sanitizing
  • 1 minute bactericidal, virucidal
Specifications
Length (English)6 in.
Length (Metric)15.2 cm
Width (English)7 in.
Width (Metric)17.8 cm
MaterialMeltblown Polypropylene
Dimensions (L x W)6 x 7 in.
Packaging Quantity12 x 160-Count Canister
Saturation SolutionPREempt RTU Disinfectant Solution
Unit SizeCase