Tool for use with the CleanSpace™ HALO Respirator unit (CS3000). Seals all the openings on the CleanSpace™ HALO PAPR unit for storage. Made of polycarbonate and silicone.
Have Questions?
Catalog number CSECS3011
Price (MYR)
400.00
EA
Add to cart
Price (MYR)
400.00
EA
Add to cart
Used with the revolutionary CleanSpace HALO PAPR: light weight, no hoses or belts
Designed for comfort over long periods
Allows sweating and breathability
Easy to wash and quick drying
Designed for long wear in harsh environments
Easy and quick replacement
Specifications
ModelCS3011
For Use With (Equipment)CleanSpace™ HALO Respirator unit (CS3000)