Costar™ Disposable Traditional Style Polystyrene Storage Bottles
Actual product may vary
Corning
Costar™ Disposable Traditional Style Polystyrene Storage Bottles
Manufactured from clear polystyrene for strength and optimum visibility. Corning™ Costar™ Disposable Storage Bottles offer the ideal solution for storing media, buffers, and other aqueous solutions
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
CRG8393500 mL
CRG8390250 mL
Catalog number CRG8393
Price (MYR)
366.00
Case
Capacity (Metric):
500 mL
Price (MYR)
366.00
Case
  • Disposable polystyrene bottles for storage of media, buffers, and other aqueous solutions
  • Costar™ traditional style has smooth sides
  • Plug seal caps (45 mm) provide an airtight seal and help minimize the risk of contamination.
  • Bottles can be used with Corning vacuum filtration systems with 45 mm neck sizes (Cat. Nos. 430626, 430627, 431161, 430049, 430513, 430514,431118, 430015, and 431174).
  • Sterile
  • Nonpyrogenic
Specifications
Capacity17 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)17 oz.
ColorClear
MaterialPolystyrene
No. per Case12/Cs.
No. per Pack1/Pk.
SterilitySterile
Closure ColorRed
Neck Finish45
ShapeCylindrical
Closure MaterialHDPE
Closure Size45 mm
Closure TypePlug Seal
Unit SizeCase