Temperature/Power Monitors
Actual product may vary
Temperature/Power Monitors
Temperature/Power Monitors
Corning
Temperature/Power Monitors
Monitor freezer performance simply, accurately and economically with this self-contained temperature/power monitor.
Have Questions?
Catalog NumberProbe TypeNo. of Probes
CRG690220 ft. (6.1m) PTFE4
Catalog number CRG6902
Price (MYR)
4,195.00
Case
Probe Type:
20 ft. (6.1m) PTFE
No. of Probes:
4
Price (MYR)
4,195.00
Case
Monitoring freezer performance simply, effectively and economically with Thermo Scientific™ Temperature/Power Monitors, which provide both alarm/monitoring and digital temperature display in a single, self-contained unit.
Features:
  • Probe size: 1.25L × 0.25 in. dia. (3.2 × 0.6cm)
  • Solid-state probes
  • High-temperature alarm with adjustable setpoint
  • Alarm mute and chirp
  • Audio delay timer
  • Remote alarm contacts (SPDT)
  • Battery backup
  • Low-voltage AC power supplys
Single-probe Digital Monitors:
  • 1 input channel
  • 1°C display and setpoint resolution
  • High-temperature alarm
  • Numeric-only LCD alphanumeric display
  • Red visual alarm
  • 8′ PTFE solid-state probe lead wire (24 gauge on 6899 model)
  • Velcro™ mounting method
  • Case dimensions: 5L × 4.5W × 2 in.D (27 × 114 × 50mm)
Multiprobe Microprocessor-based Monitors:
  • 2 input channels (6900 model), 3 input channels (6901 model) or 4 input channels (6902 model)
  • 0.1°C display and setpoint resolution
  • High- and low-temperature alarm
  • 8-character, back-lit, adjustable LCD alphanumeric display
  • Visual alarm (Green = normal; Red = deviation)
  • 20′ PTFE color-coded, solid-state probe lead wire (24 gauge on 6899 model)
  • Standard alarm/battery test switch
  • Bracket mounting method
  • Case dimensions: 6.2L × 3.25W × 2 in.D (158 × 83 × 50mm)
Specifications
TypeTemperature/Power Monitors
Temperature Range-100°C to +25°C
Door StyleSolid
For Use With (Equipment)Monitoring Freezer Temperature
Probe Type20 ft. (6.1m) PTFE
No. of Probes4
Monitoring OptionsAlarm
Height (Metric) Exterior5 cm
Length (Metric) Exterior15.9 cm
Width (Metric) Exterior8.2 cm
Unit SizeCase