Reusable Glass Spinner Flasks
Actual product may vary
Corning
Reusable Glass Spinner Flasks
High-performance
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric) OuterDiameter (Metric) Center Neck
CRG4500-500125 mL70 mm7 cm
Catalog number CRG4500-500
Price (MYR)
2,228.00
Case
Capacity (Metric):
125 mL
Diameter (Metric) Outer:
70 mm
Diameter (Metric) Center Neck:
7 cm
Price (MYR)
2,228.00
Case
  • Baffles enhance aeration and agitation of contents of the flask.
  • Unique impeller design ensures optimal stirring.
  • Sidearm designs permit easy access of 25 mL and 50 mL pipets.
Specifications
Closure ColorOrange
Closure MaterialPolypropylene
Diameter (English) Outer2.7 in.
Diameter (Metric) Center Cap7 cm
Diameter (Metric) Center Neck7 cm
Diameter (Metric) Inner Side Arm2.4 cm
Diameter (Metric) Side Arm3.2 cm
Diameter (Metric) Side Arm Cap3.2 cm
Capacity (Metric)125 mL
MaterialBorosilicate Glass
For Use With (Application)Suspension Culture
For Use With (Equipment)Magnetic Stirrer
Height (English)6.8 in.
Height (Metric)17.5 cm
IncludesFlat center cap and 2 angled side arms
Neck StyleAngled
No. per Case1/Cs.
No. per Pack1/Pk.
Product TypeSpinner Flask
Side Arm Angle45°
Side Arm Openings2
Thread SizeGL 32
AutoclavableAutoclavable
GraduatedNo
SterilityNon-sterile
Bottom ShapeFlat
Closure TypeScrew Cap
Diameter (Metric) Outer70 mm
Quantity1 Ea.
Unit SizeCase