Internally Threaded Cryogenic Vials
Actual product may vary
Internally Threaded Cryogenic Vials
Internally Threaded Cryogenic Vials
Corning
Internally Threaded Cryogenic Vials
Manufactured from polypropylene for stability in extreme temperature conditions. Corning™ Internally Threaded Cryogenic Vials provide a safe and secure vessel for ultra-low temperature storage of valuable cell cultures and biological samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Height (Metric)
CRG4304925 mL90.2 mm
CRG4306565 mL92.1 mm
Catalog number CRG430492
Price (MYR)
1,724.00
Case
Volume (Metric):
5 mL
Height (Metric):
90.2 mm
Price (MYR)
1,724.00
Case
These vials provide a safe and secure vessel for ultra-low temperature storage of valuable cell cultures and biological samples. Made of polypropylene for stability in extreme temperature conditions.
  • Manufactured from polypropylene to withstand temperatures down to -196°C
  • Black graduations with large marking spot
  • Vials have a silicone washer for a secure seal.
  • 2 mL self-standing vials are available in five cap colors.
  • Vials may be color coded with inserts.
  • Self-standing vials have a special base design allowing them to be locked into cryogenic rack and tray (Cat. No. 431131) for single-handed manipulation.
  • Free foam rack with each case
  • RNase-/DNase-free
  • Sterile
  • Nonpyrogenic
Warning! Do not use cryogenic vials for storage in the liquid phase of liquid nitrogen. Only store vials in the vapor phase above the liquefied gas. Always use appropriate safety equipment when removing vials from cryogenic storage.
Specifications
Closure ColorOrange
ColorNatural
Diameter (Metric) Outer12.7 mm
MaterialPolypropylene
SterilitySterile
Volume (Metric)5 mL
GraduatedYes
IncludesSilicone washer for secure seal, 50-position polyfoam rack with each case for easy carrying
No. per Case500
No. per Pack50
StyleRound
Temperature Range (Metric)-196°C
Thread StyleInternal
RNase FreeYes
Dnase FreeYes
Closure MaterialPolypropylene
Closure TypeCap
Marking SpotYes
Certifications/ComplianceNonpyrogenic, RNase/DNase-free
Height (Metric)90.2 mm
Unit SizeCase