Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Max. RCF | Closure Type |
|---|---|---|---|
| CRG430290 | 50 mL | 15,500 x g | Plug |
| FIS05-538-55 | 50 mL | 15,500 x g | Plug |
| FKS10314131 | 50 mL | 15,500 x g | Plug |
| CRG430776 | 250 mL | 6000 x g | Plug |
| FIS05-538-53 | 250 mL | 6000 x g | Plug |
| FKS10758961 | 250 mL | 6000 x g | Plug |