Stripettor™ Ultra Pipet Controller
Actual product may vary
Stripettor™ Ultra Pipet Controller
Stripettor™ Ultra Pipet Controller
Stripettor™ Ultra Pipet Controller
Stripettor™ Ultra Pipet Controller
Stripettor™ Ultra Pipet Controller
Corning
Stripettor™ Ultra Pipet Controller
A light, ergonomic, motorized pipet controller designed for use with glass and plastic serological pipets from 0.5 to 100mL
Have Questions?
Catalog number CRG4099
Price (MYR)
0.00
Each
Price (MYR)
0.00
Each
Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and various viscosities. Aspirating and dispensing speed is controlled through the finger triggers. When fully charged, the batteries enable up to 8 hours of continuous use.
  • LCD display for continuous visualization of speed, modes, and battery status
  • Additional colored nose cones to customize by application or user
  • Quiet powerful motor
  • Well positioned function buttons
  • Includes charging station (4998)
Specifications
Battery TypeAAA
TypePipette Controller
DisplayLCD
Filter Type0.2μm
For Use With (Equipment)Glass and plastic serological pipets from 0.5 to 100mL
IncludesUniversal power supply, Charging station (4998), three 0.2μm filters, three colored pieces and wall hanger
Number of Speeds2
Quantity1
Unit SizeEach