ART™ Non-Filtered Multichannel Pipette Tips
Actual product may vary
Corning
ART™ Non-Filtered Multichannel Pipette Tips
A variety of tip styles and configurations
Have Questions?
Catalog NumberSterilityFormat
CRG3880Non-sterileBulk
Catalog number CRG3880
Price (MYR)
6,693.00
Case
Sterility:
Non-sterile
Format:
Bulk
Price (MYR)
6,693.00
Case
A variety of tip styles and configurations
The HydroLogix™ (HLT) design utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipet tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced.
  • Available in blue or clear color
DNase, RNase, DNA, ATP, and Endotoxin Free
Specifications
Volume (Metric)1000 μL
FilteredNon-filtered
SterilityNon-sterile
ColorClear
FormatBulk
Product LineART
Unit SizeCase