Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Pipet Products
▸
Pipet Fillers
▸
Diamond SeroFlow Pipette Controllers
Actual product may vary
Corning
Diamond SeroFlow Pipette Controllers
This pipette controller has a comfortable, ergonomic design for hours of continuous use in the laboratory.
Have Questions?
Catalog Number
Motor Type
CRG3387
Digital
Catalog number
CRG3387
Price (MYR)
7,010.00
CS
Add to cart
Motor Type:
Digital
Price (MYR)
7,010.00
CS
Add to cart
Simple plug and play operation
Ergonomic, lightweight design helps reduce fatigue
Powerful motors for fast liquid draw
Precise control of coarse and fine speeds for speed and accuracy
Accommodates all sizes of serological pipettes, glass and plastic
0.45 μm hydrophobic filter prevents contamination
Long-life lithium ion battery for hours of use and no 'memory'
Use while charging without any wait time
Autoclavable nose cone ensures contamination-free pipetting
Specifications
Battery Type
Lithium-ion
Compatible Tips
All Serological Pipettes
Autoclavable
Not Autoclavable
Disposable
Reusable
Motor Type
Digital
Number of Speeds
6 Speeds