▸
Tubing
▸
Rubber and Silicone Tubing
▸
Traceable™ Silicone Pump Tubing
Actual product may vary
Corning
Traceable™ Silicone Pump Tubing
Ideal for use with food, pharmaceuticals, and other critical solutions
Have Questions?
Catalog Number
Diameter (English) Inner
For Use With
CRG3365
1/4 in.
Ultra Low Flow and Low Flow Variable-Speed Pump
Catalog number
CRG3365
Price (MYR)
1,937.00
Case
Add to cart
Diameter (English) Inner:
1/4 in.
For Use With:
Ultra Low Flow and Low Flow Variable-Speed Pump
Price (MYR)
1,937.00
Case
Add to cart
Ideal for use with food, pharmaceuticals, and other critical solutions
Replacement tubing for Variable-Speed Pump
Can be sterilized by autoclaving
Specifications
Autoclavable
Autoclavable
Diameter (English) Inner
1/4 in.
Diameter (Metric) Inner
6.35 mm
Material
Silicone
Overall Length (English)
25 ft.
Overall Length (Metric)
7.6 m
For Use With
Ultra Low Flow and Low Flow Variable-Speed Pump
Product Type
Pump Tubing