Check Order Status
Rubber Stopper for Butyrometer
Actual product may vary
Corning
Rubber Stopper for Butyrometer
Rubber Stopper for Butyrometer is ideal for sealing the bulbs of weighing butryometers.
Have Questions?
Catalog Number
Diameter (Metric) Bottom
CRG3290
9 mm
Catalog number
CRG3290
Price (MYR)
2,101.00
Case
Add to cart
Diameter (Metric) Bottom:
9 mm
Price (MYR)
2,101.00
Case
Add to cart
Rubber Stopper for Butyrometer is ideal for sealing the bulbs of weighing butryometers.
For sealing the bulbs of weighing butyrometers
Specifications
Length (Metric)
20 mm
Quantity
1 Ea.
Diameter (Metric) Bottom
9 mm
Diameter (Metric) Top
13 mm