Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Specialty Lab Equipment, Instruments, and Apparatuses
▸
Scoops, Spoons, and Spatulas
▸
Micro Double Spatula, 6mm
Actual product may vary
Corning
Micro Double Spatula, 6mm
Transport substances with ease. Usbeck™ Micro Double Spatula is ideal for lifting, flipping or spreading substances.
Have Questions?
Catalog Number
Length (Metric)
CRG3213
185 mm
Catalog number
CRG3213
Price (MYR)
534.00
Case
Add to cart
Length (Metric):
185 mm
Price (MYR)
534.00
Case
Add to cart
Stainless steel
Flexible
Specifications
Product Type
Double Ended Micro Spatula
Width (Metric) Blade
6 mm
Material
Stainless Steel
Length (Metric)
185 mm