PYREX™ Crystallizing Dishes
Actual product may vary
Corning
PYREX™ Crystallizing Dishes
Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
CRG3140-1902500 mL190 mm100 mm
CRG3140-100325 mL100 mm50 mm
FIS08-741D325 mL100 mm50 mm
FIS08-741F1200 mL150 mm75 mm
FIS08-741H2500 mL190 mm100 mm
Catalog number CRG3140-190
Price (MYR)
1,212.00
EA
Capacity (Metric):
2500 mL
Diameter (Metric):
190 mm
Height (Metric):
100 mm
Price (MYR)
1,212.00
EA
Reusable borosilicate glass dishes with heavy duty rims ideal for storage and crystallization
The rims on these dishes are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization (wet or dry). Ideal for storage and crystallization.
Specifications
AutoclavableAutoclavable
Bottom ShapeFlat
Capacity (Metric)2500 mL
DisposableReusable
MaterialBorosilicate Glass
ShapeRound
SterilitySterile
TypeCrystallizing Dish
For Use With (Application)Ideal for storage and crystallization
No. per Case6/Cs.
No. per Pack2/Pk.
Height (Metric)100 mm
Height (English)3.93 in.
Diameter (English)7.48 in.
Diameter (Metric)190 mm
Unit SizeEA