Reusable Polypropylene Low-Form Beakers, Graduated
Actual product may vary
Reusable Polypropylene Low-Form Beakers, Graduated
Reusable Polypropylene Low-Form Beakers, Graduated
Corning
Reusable Polypropylene Low-Form Beakers, Graduated
Translucent polypropylene beakers printed with blue graduations.
Have Questions?
Catalog NumberCapacity (Metric)
CRG1000P-150150 mL
Catalog number CRG1000P-150
Price (MYR)
355.00
Case
Capacity (Metric):
150 mL
Price (MYR)
355.00
Case
Translucent polypropylene beakers printed with blue graduations.
Specifications
ColorTranslucent
Diameter (English) Outer2.2 in.
Diameter (Metric) Outer5.6 cm
Capacity (Metric)150 mL
MaterialPolypropylene
For Use With (Application)For High Temperature and Chemical Resistance
Height (English)3.1 in.
Height (Metric)8 cm
Increments20 mL
Temperature (English) Max. Operating140°F
Temperature (Metric) Max. Operating60°C
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Certifications/ComplianceDIN, ASTM Class A and B
ScaleBlue Graduated Scale
StyleGriffin
Unit SizeCase