Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Hotplates and Stirrers
▸
Magnetic Stirrers
▸
elements™ Modular Magnetic Stirrer
Actual product may vary
Cole-Parmer
elements™ Modular Magnetic Stirrer
Optimize your stirring needs and workspace with this completely customizable stirrer
Have Questions?
Change view
Catalog Number
Type
Stirring Positions
CLP84003-82
Stirring Unit
1
CLP84003-81
Stirrer Control Unit
10
2 Options
Catalog number
CLP84003-82
Price (MYR)
-
Type:
Stirring Unit
Stirring Positions:
1
Attach up to 10 stir plates based on need
Simple touch and slide operation
LCD display with real time parameters
Corrosion-resistant glass stir top
Waterproof USB attachments
Specifications
Speed
100 to 1500 rpm
Stirring Positions
1
Top Plate Material
Glass
Capacity
1 L/position
Display
Digital
Shape
Round
Voltage
100/240 V
Top Plate Diameter (English)
3.75 in.
Frequency
50/60 Hz
Type
Stirring Unit