EasyReach™ Cleaning System: Cleaning Tool
Actual product may vary
Masterflex
EasyReach™ Cleaning System: Cleaning Tool
Passes easily through transfer hatches, requires no assembly and is maintenance-free. EasyReach™ Cleaning System is ideal for isolators, cabinets, laminar air-flow hoods, glove boxes and other mini environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialProduct TypeLength (English)
CLP40622-47Stainless SteelLay Flat Mop Frame16.5 in.
CTC2665LFStainless SteelLay Flat Mop Frame16.5 in.
Catalog number CLP40622-47
Price (MYR)
-
Material:
Stainless Steel
Product Type:
Lay Flat Mop Frame
Length (English):
16.5 in.
EasyReach™ Cleaning System consists of a lightweight stainless steel 1 piece cleaning tool and interchangeable disposable and absorbant cleaning pads that provide effective cleaning and disinfection in hard to reach areas.
  • Rectangular shape makes overlapping straight-line action easy to achieve
  • Low profile 360° pivoting head allows quick easy access to awkward areas
  • Hinged joint and lightweight design
  • Easy grab edge on the cleaning pad ensures pad fits tightly on the mop head frame; easy to replace with gloved hands
  • Small pack quantities are optimized for single session use
  • Well-fitted quilted polyester pad ensures effective, even surface coverage, reducing the number of viable and non-viable contaminants and ensuring maximum residue removal
  • Sorbent double layer Quiltec™ sealed-edge pads ensure particles and fibers are minimized
  • Cleaning pads available presaturated with 70% IPA
Note: Cleaning Pads are sold separately (see MEQT0001, MEQT0002, PSME0001). See QuickConnect™ Mop head frames and handles for long handled EasyReach option.
Specifications
Color CodeWhite
Product TypeLay Flat Mop Frame
Length (English)16.5 in.
SterilityNon-sterile
Length (Metric)42 cm
AutoclavableAutoclavable
DisposableReusable
Color-codedNo