BCR™ Cleanroom MyNotes
BCR™ Cleanroom MyNotes
Actual product may vary
Berkshire
BCR™ Cleanroom MyNotes
Our BCR™ Mynotes are made with the purest quality cellulose from specially selected forests and is specially coated to minimize particle and fiber release
Have Questions?
Catalog NumberSize
BERMNBK-A4CRA4
Catalog number BERMNBK-A4CR
Price (MYR)
-
Size:
A4
They are practical for note taking and data recording in the cleanroom. Bond is made with the purest quality cellulose from specially selected sustainable forests and is coated with a proprietary formula to minimise particle and fibre release.
Specifications
MaterialCellulose
RuledCollege Ruled
Certifications/ComplianceISO Class 3 - 7, Cleanroom Class 1 - 10,000, EU Grade B/C/D
Packaging50 Sheets/book
SizeA4
Unit SizeCase