Transfer Vessels Dipper
Actual product may vary
Transfer Vessels Dipper
Transfer Vessels Dipper
Thermo Scientific
Transfer Vessels Dipper
Remove small amounts of liquid nitrogen by using Thermo Scientific™ Transfer Vessels Dipper.
Have Questions?
Catalog number AY509X6
HSN 73269099
Price (MYR)
1,769.00
Each
Price (MYR)
1,769.00
Each
Remove small amounts of liquid nitrogen by using Thermo Scientific™ Transfer Vessels Dipper. This dipper is very convenient in the lab for liquid nitrogen storage sample preparation.
Specifications
DescriptionLN2 dipper, 12mL
TypeLN2 Vessel Dipper
Capacity (Metric)12 mL
For Use With (Equipment)LN2 Transfer Vessels
Unit SizeEach