Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
Acetonitrile (HPLC), Fisher Chemical
Thermo Scientific Chemicals
Acetonitrile (HPLC), Fisher Chemical
HPLC mobile phase for analytical and preparative separations
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Packaging:
View all
Amber Glass Bottle
Catalog number ALFA998-4
Price (MYR)
309.00
Each
Quantity:
4 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
309.00
Each
Specifications
Chemical Name or MaterialAcetonitrile
Cleanup SymbolSolvent
Melting Point-46°C
Absorbance0.01 max. at 254nm, 0.02 max. at 220nm, 0.04 max. at 210nm, 0.05 max. at 205nm, 0.07 max. at 200nm, 0.15 max. at 195nm, 1 max. at 190nm, Pass Test
CAS75-05-8
View more