3-(Bromoacetyl)coumarin, 97%, Thermo Scientific™
3-(Bromoacetyl)coumarin, 97%, Thermo Scientific™
3-(Bromoacetyl)coumarin, 97%, Thermo Scientific™
3-(Bromoacetyl)coumarin, 97%, Thermo Scientific™
Acros Organics
3-(Bromoacetyl)coumarin, 97%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ACR456550050
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3-(Bromoacetyl)coumarin
CAS29310-88-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact len