Penicillin G sodium salt, 96+%, Thermo Scientific Chemicals
Penicillin G sodium salt, 96+%, Thermo Scientific Chemicals
Penicillin G sodium salt, 96+%, Thermo Scientific Chemicals
Acros Organics
Penicillin G sodium salt, 96+%, Thermo Scientific Chemicals
Penicillin G sodium salt, CAS # 69-57-8, also called benzylpenicillin, is a β-lactam antibiotic active against gram-positive and gram-negative bacteria.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number ACR455690050
Price (MYR)
486.00
Each
Quantity:
5 g
Request bulk or custom format
Price (MYR)
486.00
Each
Specifications
Chemical Name or MaterialPenicillin G sodium salt
TypePenicillin G Sodium Salt
CAS69-57-8
Health Hazard 1Danger
pH5.0 to 7.5
View more
General Description
  • Penicillin G sodium salt is the sodium salt of benzylpenicillin, a broad-spectrum β-lactam antibiotic with gram-positive and gram-negative bactericidal activity
  • Penicillin G binds and inactivates penicillin-binding-proteins associated with the bacterial cell wall, thus inhibiting cell wall synthesis and resulting in cell lysis
Applications
  • Penicillin G can be used as a selection agent in the isolation of microorganisms or in cell culture applications
  • It can also be used to prevent infection in research animals
RUO – Research Use Only
General References:
  1. Livermore, D.M. beta-Lactamases- the Threat Renews. Curr. Protein Pept. Sci. 2009, 10, (5), 397-400.
  2. Demain, A.L. Production of beta-lactam antibiotics and its regulation. Proc. Natl. Sci. Counc. Repub. China B. 1991, 15, (4), 251-65.