Sulbactam
Sulbactam
Sulbactam
Acros Organics
Sulbactam
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR455350050
Price (MYR)
724.00
Each
Quantity:
5 g
Request bulk or custom format
Price (MYR)
724.00
Each
Specifications
Chemical Name or MaterialSulbactam
CAS68373-14-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.