4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%
Acros Organics
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%
CAS: 81-81-2 | C19H16O4 | 308.34 g/mol
Have Questions?
Catalog NumberQuantity
ACR45468010010 g
Catalog number ACR454680100
Price (MYR)
-
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin
CAS81-81-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
Health Hazard 3GHS P Statement
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.