Potassium Chloride, 99+%, ACS Reagent
Potassium Chloride, 99+%, ACS Reagent
Potassium Chloride, 99+%, ACS Reagent
Thermo Scientific Chemicals
Potassium Chloride, 99+%, ACS Reagent
CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR424090010
Price (MYR)
878.00
Bottle
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
878.00
Bottle
Specifications
Chemical Name or MaterialPotassium Chloride
CAS7447-40-7
ColorWhite
Health Hazard 1HYGROSCOPIC
pH5.4 to 8.6 (5% soln. at 25°C)