Silver nitrate, 99+%, ACS reagent
Silver nitrate, 99+%, ACS reagent
Silver nitrate, 99+%, ACS reagent
Thermo Scientific Chemicals
Silver nitrate, 99+%, ACS reagent
CAS: 7761-88-8 | AgNO3 | 169.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR419350500
Price (MYR)
3,176.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,176.00
Each
Specifications
Boiling Point444.0°C
Chemical Name or MaterialSilver nitrate
Melting Point212.0°C
CAS7761-88-8