▸
Organic compounds
▸
Benzenoids
▸
Benzene and substituted derivatives
▸
Benzoyl derivatives
▸
3-chloro-5-fluorobenzaldehyde, 97%, Thermo Scientific™
Element Search
Structure Search
Acros Organics
3-chloro-5-fluorobenzaldehyde, 97%, Thermo Scientific™
Have Questions?
Change view
Quantity:
20 g
5 g
Packaging:
Glass bottle
Catalog number
ACR374750200
Price (MYR)
-
Quantity:
20 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
3-chloro-5-fluorobenzaldehyde
CAS
90390-49-1
CAS Min %
96.0
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0