Thiophene-2-ethylamine, 98%
Thiophene-2-ethylamine, 98%
Thiophene-2-ethylamine, 98%
Acros Organics
Thiophene-2-ethylamine, 98%
CAS: 30433-91-1 | C6H9NS | 127.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR354270050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialThiophene-2-ethylamine
CAS30433-91-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre