Lithium hydride, 97%, pure, powder (<100 μm)
Lithium hydride, 97%, pure, powder (<100 μm)
Lithium hydride, 97%, pure, powder (<100 μm)
Acros Organics
Lithium hydride, 97%, pure, powder (<100 μm)
CAS: 7580-67-8 | HLi | 7.95 g/mol
Have Questions?
Catalog NumberQuantity
ACR332955000500 g
Catalog number ACR332955000
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium hydride
Melting Point680.0°C
Name Note98%
CAS7580-67-8
ColorWhite to Gray
View more
Reduction reagent
RUO – Research Use Only