2,6-Difluorobenzaldehyde, 98%
2,6-Difluorobenzaldehyde, 98%
2,6-Difluorobenzaldehyde, 98%
Acros Organics
2,6-Difluorobenzaldehyde, 98%
CAS: 437-81-0 | C7H4F2O | 142.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR311460050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 6-Difluorobenzaldehyde
CAS437-81-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0